Assay Details
| Gene Symbol: LINC00559 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_047489.1 | 1 - 2 | 946 | 68 | ||
| GenBank mRNA | BC043442.1 | - | 1 - 2 | 950 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 353 | LINC00353 | -542.2kb | Sense | Chr. 13 - 89548794 - 89564413 on NCBI Build GRCh38 | |
| Downstream | microRNA 622 | MIR622 |
MIRN622, hsa-mir-622 |
111.5kb | Anti-sense | Chr. 13 - 90231182 - 90231277 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163239.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163239.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163239.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163239.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163239.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163239.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163239.hg.1 | TC1300009443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300009443.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163241.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163241.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163241.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163241.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163241.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163241.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163241.hg.1 | TC1300009443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300163240.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163240.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163240.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163240.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163240.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163240.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163240.hg.1 | TC1300009443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300163242.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163242.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163242.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163242.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163242.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163242.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163242.hg.1 | TC1300009443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300163238.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163238.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163238.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163238.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163238.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163238.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163238.hg.1 | TC1300009443.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300163244.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163244.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163244.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163244.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163244.hg.1 | ENST00000569476.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300163244.hg.1 | NR_047489.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300163244.hg.1 | TC1300009443.hg.1 | Clariom D TC |

LINC00559
Genomic Map