Assay Details
| Gene Symbol: LOC101927363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134672.1 | 1 - 2 | 110 | 122 | ||
| GenBank mRNA | BC043520.1 | - | 1 - 2 | 110 | 122 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723828 | LOC102723828 | -195.6kb | Sense | Chr. 4 - 31997379 - 32155406 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928622 | LOC101928622 | 1.5mb | Anti-sense | Chr. 4 - 33896339 - 34039893 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101927363
Genomic Map