Assay Details
| Gene Symbol: LOC101927693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC044238.1 | - | 1 - 2 | 223 | 103 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BM908133.1 | - | 233 | Chr. - - [-] on Build | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | meiotic kinetochore factor | MEIKIN | -57.9kb | Anti-sense | Chr. 5 - 131806990 - 131945698 on NCBI Build GRCh38 | |
| Downstream | interleukin 3 | IL3 |
IL-3, MCGF, MULTI-CSF |
53.6kb | Sense | Chr. 5 - 132060654 - 132063203 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101927693
Genomic Map