Assay Details
| Gene Symbol: LOC101929159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135550.1 | 1 - 2 | 139 | 105 | ||
| GenBank mRNA | BC044922.2 | - | 1 - 2 | 139 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DENN domain containing 6A | DENND6A |
AFI1A, FAM116A |
-116.0 | Anti-sense | Chr. 3 - 57625454 - 57693118 on NCBI Build GRCh38 |
| Downstream | sarcolemma associated protein | SLMAP | SLAP | 59.6kb | Sense | Chr. 3 - 57756221 - 57930013 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101929159
Genomic Map