Assay Details
| Gene Symbol: LOC101928992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110043.1 | 2 - 3 | 330 | 60 | ||
| GenBank mRNA | BC050282.1 | - | 2 - 3 | 330 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MAGE family member F1 | MAGEF1 | -44.3kb | Sense | Chr. 3 - 184710367 - 184712048 on NCBI Build GRCh38 | |
| Downstream |
VPS8, CORVET complex subunit |
VPS8 | KIAA0804 | 46.7kb | Anti-sense | Chr. 3 - 184812137 - 185052614 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000397759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000397759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000397759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000595703.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000595703.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000595703.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000595703.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000595703.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000596883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000596883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000596883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000596883.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000601686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000601686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000601686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000601686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | ENST00000601686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199906.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199906.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199906.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199906.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199906.hg.1 | TC0300013401.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300199905.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199905.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199905.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199905.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199905.hg.1 | TC0300013401.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013401.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199904.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199904.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199904.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199904.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199904.hg.1 | TC0300013401.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000397755.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000397759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000397759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000397759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000430298.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000595703.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000595703.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000595703.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000595703.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000595703.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000596096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000596883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000596883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000596883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000596883.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000601686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000601686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000601686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000601686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | ENST00000601686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007063.hg.1 | NR_027049.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199903.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199903.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199903.hg.1 | ENST00000457449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300199903.hg.1 | NR_110043.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300199903.hg.1 | TC0300013401.hg.1 | Clariom D TC |

LOC101928992
Genomic Map