Assay Details
| Gene Symbol: LOC101927598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC055421.1 | - | 1 - 2 | 1873 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | FERM domain containing 6 | FRMD6 |
C14orf31, EX1, Willin, c14_5320 |
-34.5kb | Sense | Chr. 14 - 51489100 - 51730727 on NCBI Build GRCh38 |
| Downstream | G protein subunit gamma 2 | GNG2 | 57.3kb | Sense | Chr. 14 - 51847131 - 51969800 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000450217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000450217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000450217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000450217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000450217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000450217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000450217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000450217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000450217.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526284.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000526488.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000529482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000530366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000530366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000530366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000530366.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | ENST00000533775.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012952.hg.1 | NR_024248.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007152.hg.1 | ENST00000553312.1 | Ensembl |

LOC101927598
Genomic Map