Assay Details
| Gene Symbol: WLS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX537492.1 | - | 1 | 7247 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GNG12 antisense RNA 1 | GNG12-AS1 | Anti-sense | Chr. 1 - 67832288 - 68202987 on NCBI Build GRCh38 | ||
| Downstream | microRNA 1262 | MIR1262 |
MIRN1262, hsa-mir-1262, mir-1262 |
77.1kb | Sense | Chr. 1 - 68183518 - 68183610 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000414904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000414904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000414904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000414904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000434072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | ENST00000434072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000413628.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000414904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000414904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000414904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000414904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000420587.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000434072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | ENST00000434072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008667.hg.1 | NR_040077.1 | Refseq |

WLS
Genomic Map