Assay Details
| Gene Symbol: LOC105373414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX538340.1 | - | 1 | 3148 | 92 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BM981387.1 | - | 1 | Chr. 2 - 9109143 - 9109870 [-] on Build GRCh38 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | membrane bound O-acyltransferase domain containing 2 | MBOAT2 |
LPAAT, LPCAT4, LPEAT, LPLAT 2, OACT2 |
-102.8kb | Anti-sense | Chr. 2 - 8852690 - 9003750 on NCBI Build GRCh38 |
| Downstream |
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2 |
ASAP2 |
AMAP2, CENTB3, DDEF2, PAG3, PAP, Pap-alpha, SHAG1 |
96.9kb | Sense | Chr. 2 - 9206765 - 9405683 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105373414
Genomic Map