Assay Details
| Gene Symbol: LSAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011512840.2 | XP_011511142.1 | 8 | 4529 | 80 | |
| GenBank mRNA | BX647284.1 | - | 1 | 5149 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | growth associated protein 43 | GAP43 |
B-50, PP46 |
-106.8kb | Anti-sense | Chr. 3 - 115623304 - 115721487 on NCBI Build GRCh38 |
| Downstream | LSAMP antisense RNA 1 | LSAMP-AS1 | 526.4kb | Anti-sense | Chr. 3 - 116360024 - 116370085 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LSAMP
Genomic Map