Assay Details
| Gene Symbol: LINC01450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110831.1 | 2 - 3 | 633 | 131 | ||
| GenBank mRNA | BX647937.1 | - | 2 - 3 | 634 | 131 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | succinyl-CoA:glutarate-CoA transferase | SUGCT |
C7orf10, DERP13, GA3, ORF19 |
-103.9kb | Anti-sense | Chr. 7 - 40134887 - 40860767 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1449 | LINC01449 | 133.4kb | Anti-sense | Chr. 7 - 41101604 - 41133501 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01450
Genomic Map