Assay Details
| Gene Symbol: LRRC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017001888.1 | XP_016857377.1 | 26 | 5825 | 90 | |
| GenBank mRNA | BX648943.1 | - | 1 | 7457 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
peptidylprolyl cis/trans isomerase, NIMA-interacting 1 pseudogene 1 |
PIN1P1 | PIN1L | -168.7kb | Sense | Chr. 1 - 69919322 - 69920317 on NCBI Build GRCh38 |
| Downstream | leucine rich repeat containing 40 | LRRC40 | dJ677H15.1 | 48.2kb | Anti-sense | Chr. 1 - 70144802 - 70205678 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LRRC7
Genomic Map