Assay Details
| Gene Symbol: LINC01043 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135321.1 | 6250 | 89 | |||
| GenBank mRNA | BX649107.1 | - | 6250 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928730 | LOC101928730 | -46.0kb | Anti-sense | Chr. 13 - 112256725 - 112273000 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1044 | LINC01044 | 1.4kb | Anti-sense | Chr. 13 - 112322567 - 112331225 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC01043
Genomic Map