Assay Details
| Gene Symbol: NR3C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123047.1 | - | 1 | 1546 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho GTPase activating protein 10 | ARHGAP10 |
GRAF2, PS-GAP, PSGAP |
-186.8kb | Anti-sense | Chr. 4 - 147732063 - 148072781 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105377480 | LOC105377480 | 678.8kb | Sense | Chr. 4 - 148940946 - 148942378 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NR3C2
Genomic Map