Assay Details
| Gene Symbol: CA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123309.1 | - | 1 | 650 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | CA3 antisense RNA 1 | CA3-AS1 | -14.3kb | Anti-sense | Chr. 8 - 85441806 - 85464915 on NCBI Build GRCh38 | |
| Downstream |
REX1, RNA exonuclease 1 homolog-like 1, pseudogene |
REXO1L1P |
GOR, REXO1L1 |
180.0kb | Anti-sense | Chr. 8 - 85661470 - 85663497 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

CA2
Genomic Map