Assay Details
Gene Symbol: GRM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK123326.1 | - | 1 | 1751 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | GRM7 antisense RNA 1 | GRM7-AS1 | -203.3kb | Anti-sense | Chr. 3 - 7519741 - 7535284 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC101927394 | LOC101927394 | 211.3kb | Anti-sense | Chr. 3 - 7952805 - 8016307 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
