| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006478.hg.1 |
ENST00000551144.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006478.hg.1 |
ENST00000551144.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006478.hg.1 |
ENST00000551144.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006478.hg.1 |
ENST00000611741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006478.hg.1 |
ENST00000611741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400006478.hg.1 |
ENST00000611741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011672.hg.1 |
ENST00000491608.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011672.hg.1 |
ENST00000491608.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011672.hg.1 |
ENST00000491608.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011672.hg.1 |
ENST00000491608.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011672.hg.1 |
ENST00000491608.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011672.hg.1 |
ENST00000503597.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400011672.hg.1 |
ENST00000503597.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007855.hg.1 |
ENST00000428118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007855.hg.1 |
ENST00000428118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007855.hg.1 |
ENST00000428118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009268.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009268.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009268.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009268.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009268.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009268.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009268.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200009268.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007855.hg.1 |
ENST00000428118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007855.hg.1 |
ENST00000428118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007855.hg.1 |
ENST00000428118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009268.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009268.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009268.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009268.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200009268.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009268.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009268.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200009268.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400006478.hg.1 |
ENST00000551144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006478.hg.1 |
ENST00000551144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006478.hg.1 |
ENST00000551144.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006478.hg.1 |
ENST00000611741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006478.hg.1 |
ENST00000611741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400006478.hg.1 |
ENST00000611741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200164862.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200164862.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200164862.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200164862.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200164862.hg.1 |
TC0200009268.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400011672.hg.1 |
ENST00000491608.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011672.hg.1 |
ENST00000491608.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011672.hg.1 |
ENST00000491608.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011672.hg.1 |
ENST00000491608.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011672.hg.1 |
ENST00000491608.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011672.hg.1 |
ENST00000503597.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400011672.hg.1 |
ENST00000503597.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200164863.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200164863.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200164863.hg.1 |
ENST00000450840.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200164863.hg.1 |
NR_110285.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200164863.hg.1 |
TC0200009268.hg.1 |
Clariom D TC |