Assay Details
| Gene Symbol: ZDHHC14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123429.1 | - | 1 | 1052 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3692 | MIR3692 | -145.7kb | Sense | Chr. 6 - 157529132 - 157529200 on NCBI Build GRCh38 | |
| Downstream | sorting nexin 9 | SNX9 |
SDP1, SH3PX1, SH3PXD3A, WISP |
38.8kb | Sense | Chr. 6 - 157716046 - 157945077 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZDHHC14
Genomic Map