Assay Details
| Gene Symbol: TBATA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124147.1 | - | 1 | 1453 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADAM metallopeptidase with thrombospondin type 1 motif 14 | ADAMTS14 | -8.8kb | Anti-sense | Chr. 10 - 70672518 - 70762441 on NCBI Build GRCh38 | |
| Downstream | sphingosine-1-phosphate lyase 1 | SGPL1 |
S1PL, SPL |
42.0kb | Anti-sense | Chr. 10 - 70815920 - 70881189 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000394982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000394982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000394982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000394982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000394982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000299290.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000394982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000394982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000394982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000394982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000394982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | ENST00000456372.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010926.hg.1 | NM_152710.3 | Refseq |

TBATA
Genomic Map