Assay Details
| Gene Symbol: TMTC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124599.1 | - | 1 | 2651 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gamma-glutamylamine cyclotransferase | GGACT | A2LD1 | -15.1kb | Sense | Chr. 13 - 100530164 - 100588792 on NCBI Build GRCh38 |
| Downstream | NALCN antisense RNA 1 | NALCN-AS1 | 100.2kb | Anti-sense | Chr. 13 - 100708325 - 101059286 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TMTC4
Genomic Map