| Array Name | 
									Probe IDs | 
									Targets | 
									Target Type | 
								
	
								
									
										
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Transcriptome Array 2.0 | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169745.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169745.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169745.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169745.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169745.hg.1 | 
												TC1200009358.hg.1 | 
												Clariom D TC | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												TC1200009358.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169747.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169747.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169747.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169747.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169747.hg.1 | 
												TC1200009358.hg.1 | 
												Clariom D TC | 
												
											
										
										
									
								
									
										
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169746.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169746.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169746.hg.1 | 
												ENST00000541258.1 | 
												Ensembl | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169746.hg.1 | 
												NR_104648.1 | 
												Refseq | 
												
											
												
												| Affymetrix Human Clariom D Assay | 
												PSR1200169746.hg.1 | 
												TC1200009358.hg.1 | 
												Clariom D TC |