Assay Details
| Gene Symbol: FBXO32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242463.1 | NP_001229392.1 | 7 | 2429 | 88 | |
| NM_058229.3 | NP_478136.1 | 9 | 2708 | 88 | ||
| NM_148177.2 | NP_680482.1 | 6 | 2343 | 88 | ||
| GenBank mRNA | AK125888.1 | - | 1 | 847 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WDYHV motif containing 1 | WDYHV1 | C8orf32 | -49.5kb | Anti-sense | Chr. 8 - 123416725 - 123467527 on NCBI Build GRCh38 |
| Downstream | kelch like family member 38 | KLHL38 | C8ORFK36 | 142.7kb | Sense | Chr. 8 - 123644442 - 123653829 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179332.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179332.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179332.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179332.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179332.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179332.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179332.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179332.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179332.hg.1 | TC0800011683.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000287396.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000443022.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000517956.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000520511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000521719.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | ENST00000524000.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_001242463.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_058229.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011683.hg.1 | NM_148177.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 133754 | - | - |

FBXO32
Genomic Map