Assay Details
| Gene Symbol: AKAP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126161.1 | - | 1 | 2754 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723831 | LOC102723831 | -95.1kb | Anti-sense | Chr. 6 - 151196673 - 151228457 on NCBI Build GRCh38 | |
| Downstream | zinc finger and BTB domain containing 2 | ZBTB2 | ZNF437 | 37.6kb | Anti-sense | Chr. 6 - 151364115 - 151391705 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

AKAP12
Genomic Map