Assay Details
| Gene Symbol: LOC105373326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK126232.1 | - | 1 | 2641 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ATPase Na+/K+ transporting family member beta 4 | ATP1B4 | -47.7kb | Anti-sense | Chr. X - 120362085 - 120383249 on NCBI Build GRCh38 | |
| Downstream | cullin 4B | CUL4B |
CUL-4B, MRXHF2, MRXS15, MRXSC, SFM2 |
90.4kb | Sense | Chr. X - 120524589 - 120575829 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105373326
Genomic Map