| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010186.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010186.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010186.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010186.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010186.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010186.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010186.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010186.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010186.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010186.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900183792.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900183792.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900183792.hg.1 |
TC1900010186.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900183792.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900183792.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900183791.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900183791.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900183791.hg.1 |
TC1900010186.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900183791.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900183791.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010186.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010186.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010186.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010186.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010186.hg.1 |
ENST00000562262.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010186.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010186.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010186.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010186.hg.1 |
NR_110427.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010186.hg.1 |
NR_110427.1 |
Refseq |