Assay Details
| Gene Symbol: COG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161520.1 | NP_001154992.1 | 21 | 5542 | 91 | |
| GenBank mRNA | AK126275.1 | - | 1 | 2347 | 91 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HMG-box transcription factor 1 | HBP1 | -5.3kb | Anti-sense | Chr. 7 - 107168961 - 107202529 on NCBI Build GRCh38 | |
| Downstream | G protein-coupled receptor 22 | GPR22 | 259.7kb | Anti-sense | Chr. 7 - 107470057 - 107476207 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

COG5
Genomic Map