Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ADAMTS9 antisense RNA 2 | ADAMTS9-AS2 | -556.9kb | Anti-sense | Chr. 3 - 64684870 - 65011468 on NCBI Build GRCh38 | |
| Downstream | MAGI1 antisense RNA 1 | MAGI1-AS1 | 323.7kb | Anti-sense | Chr. 3 - 65893816 - 65925297 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map