Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4251 | MIR4251 | -78.0kb | Sense | Chr. 1 - 3127975 - 3128035 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105378604 | LOC105378604 | 98.0kb | Anti-sense | Chr. 1 - 3306636 - 3327974 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map