Assay Details
| Gene Symbol: EBF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017009192.1 | XP_016864681.1 | 16 | 4790 | 82 | |
| XM_017009193.1 | XP_016864682.1 | 16 | 4775 | 82 | ||
| XM_017009194.1 | XP_016864683.1 | 16 | 4766 | 82 | ||
| XM_017009195.1 | XP_016864684.1 | 16 | 4763 | 82 | ||
| XM_017009196.1 | XP_016864685.1 | 16 | 5725 | 82 | ||
| XM_017009197.1 | XP_016864686.1 | 16 | 5228 | 82 | ||
| XM_017009199.1 | XP_016864688.1 | 11 | 5141 | 82 | ||
| XM_017009200.1 | XP_016864689.1 | 11 | 22277 | 82 | ||
| XM_017009204.1 | XP_016864693.1 | 8 | 4667 | 82 | ||
| GenBank mRNA | AK130895.1 | - | 1 | 2152 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927697 | LOC101927697 | -294.9kb | Sense | Chr. 5 - 158320683 - 158409773 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101927740 | LOC101927740 | 393.5kb | Anti-sense | Chr. 5 - 159100483 - 159117478 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

EBF1
Genomic Map