Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NACHT and WD repeat domain containing 2 | NWD2 | KIAA1239 | -75.7kb | Sense | Chr. 4 - 37245068 - 37449465 on NCBI Build GRCh38 |
| Downstream | RELT like 1 | RELL1 | 60.9kb | Anti-sense | Chr. 4 - 37590800 - 37686377 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map