Assay Details
Gene Symbol: LOC105373942 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK026429.1 | - | 1 | 2951 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | AGAP1 intronic transcript 1 | AGAP1-IT1 | -238.8kb | Anti-sense | Chr. 2 - 235505751 - 235507566 on NCBI Build GRCh38 | |
Downstream | translation initiation factor IF-2-like | LOC105373944 | 412.0kb | Sense | Chr. 2 - 236161340 - 236164886 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
