Assay Details
| Gene Symbol: GPR20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK026750.1 | - | 1 | 1849 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1300 | LINC01300 | -8.8kb | Anti-sense | Chr. 8 - 141340549 - 141344621 on NCBI Build GRCh38 | |
| Downstream |
protein tyrosine phosphatase type IVA, member 3 |
PTP4A3 |
PRL-3, PRL-R, PRL3 |
36.6kb | Anti-sense | Chr. 8 - 141391993 - 141432454 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GPR20
Genomic Map