Assay Details
| Gene Symbol: FAM78B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320302.1 | NP_001307231.1 | 2 | 2509 | 92 | |
| NR_135199.1 | 3 | 3441 | 92 | |||
| XM_017000417.1 | XP_016855906.1 | 3 | 7647 | 92 | ||
| XM_017000418.1 | XP_016855907.1 | 3 | 2989 | 92 | ||
| GenBank mRNA | AB593134.1 | - | 2 | 2509 | 92 | |
| AK055347.1 | - | 1 | 548 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uridine-cytidine kinase 2 | UCK2 |
TSA903, UK, UMPK |
-145.8kb | Anti-sense | Chr. 1 - 165827495 - 165911618 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105371586 | LOC105371586 | 21.4kb | Anti-sense | Chr. 1 - 166060611 - 166091339 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

FAM78B
Genomic Map