Assay Details
| Gene Symbol: SLC6A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317406.1 | NP_001304335.1 | 4 | 10229 | 72 | |
| GenBank mRNA | AK055416.1 | - | 1 | 1248 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 606 | LINC00606 | -67.5kb | Anti-sense | Chr. 3 - 10759484 - 10764192 on NCBI Build GRCh38 | |
| Downstream | solute carrier family 6 member 1 | SLC6A1 |
GABATHG, GABATR, GAT1, MAE |
158.9kb | Sense | Chr. 3 - 10992718 - 11039249 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SLC6A11
Genomic Map