Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SNRK antisense RNA 1 | SNRK-AS1 | -16.7kb | Sense | Chr. 3 - 43346919 - 43351962 on NCBI Build GRCh38 | |
| Downstream | abhydrolase domain containing 5 | ABHD5 |
CDS, CGI58, IECN2, NCIE2 |
320.0kb | Anti-sense | Chr. 3 - 43690883 - 43722725 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map