Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ciliary rootlet coiled-coil, rootletin family member 2 |
CROCC2 | -82.8kb | Sense | Chr. 2 - 240919048 - 240993311 on NCBI Build GRCh38 | |
| Downstream | PAS domain containing serine/threonine kinase | PASK |
PASKIN, STK37 |
27.7kb | Anti-sense | Chr. 2 - 241106099 - 241150347 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map