Assay Details
| Gene Symbol: LINC01310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038944.1 | 1 - 2 | 80 | 62 | ||
| GenBank mRNA | AK056844.1 | - | 1 - 2 | 80 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4535 | MIR4535 | -86.4kb | Sense | Chr. 22 - 48780295 - 48780353 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105373085 | LOC105373085 | 162.4kb | Anti-sense | Chr. 22 - 49047480 - 49052405 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000409.hg.1
Set Membership: |
|

LINC01310
Genomic Map