Assay Details
| Gene Symbol: FEZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057492.1 | - | 1 | 845 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cysteine rich transmembrane BMP regulator 1 | CRIM1 |
CRIM-1, S52 |
-1.1kb | Anti-sense | Chr. 2 - 36356088 - 36551135 on NCBI Build GRCh38 |
| Downstream | vitrin | VIT | 142.4kb | Anti-sense | Chr. 2 - 36696685 - 36814794 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

FEZ2
Genomic Map