Assay Details
| Gene Symbol: LINC01547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127059.1 | - | 1 | 3556 | 63 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ITGB2 antisense RNA 1 | ITGB2-AS1 | -6.3kb | Anti-sense | Chr. 21 - 44921035 - 44929680 on NCBI Build GRCh38 | |
| Downstream | family with sequence similarity 207 member A | FAM207A |
C21orf70, PRED56 |
85.0 | Anti-sense | Chr. 21 - 44939697 - 44976989 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LINC01547
Genomic Map