Assay Details
| Gene Symbol: DIDO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011528506.1 | XP_011526808.1 | 1 | 3048 | 70 | |
| GenBank mRNA | AK127121.1 | - | 1 | 3048 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transcription factor like 5 | TCFL5 |
CHA, E2BP-1, Figlb, bHLHe82 |
-68.0kb | Sense | Chr. 20 - 62841014 - 62861763 on NCBI Build GRCh38 |
| Downstream | GID complex subunit 8 homolog | GID8 |
C20orf11, TWA1 |
4.7kb | Anti-sense | Chr. 20 - 62938089 - 62948475 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011528506.1 transcripts.
Set Membership: |
|

DIDO1
Genomic Map