Assay Details
| Gene Symbol: LOC105371307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127458.1 | - | 1 | 355 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cadherin 8 | CDH8 | Nbla04261 | -1.0mb | Sense | Chr. 16 - 61640435 - 62036835 on NCBI Build GRCh38 |
| Downstream | cadherin 11 | CDH11 |
CAD11, CDHOB, OB, OSF-4 |
1.9mb | Sense | Chr. 16 - 64943753 - 65126112 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105371307
Genomic Map