Assay Details
| Gene Symbol: LOC101928272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120635.1 | 1 - 2 | 45 | 94 | ||
| GenBank mRNA | AK127590.1 | - | 1 - 2 | 45 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 709 | LINC00709 | -7.3kb | Anti-sense | Chr. 10 - 9275613 - 9287057 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928298 | LOC101928298 | 763.1kb | Anti-sense | Chr. 10 - 10058722 - 10063502 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928272
Genomic Map