Assay Details
| Gene Symbol: LOC101928272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120635.1 | 3 - 4 | 420 | 64 | ||
| GenBank mRNA | AK127590.1 | - | 3 - 4 | 420 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105376398 | LOC105376398 | -675.9kb | Sense | Chr. 10 - 8572747 - 8594391 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928298 | LOC101928298 | 774.3kb | Anti-sense | Chr. 10 - 10058722 - 10063502 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928272
Genomic Map