Assay Details
| Gene Symbol: LOC101928272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120635.1 | 4 - 5 | 596 | 84 | ||
| GenBank mRNA | AK127590.1 | - | 4 - 5 | 596 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105376398 | LOC105376398 | -603.0kb | Sense | Chr. 10 - 8572747 - 8594391 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 709 | LINC00709 | 5.2kb | Anti-sense | Chr. 10 - 9275613 - 9287057 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928272
Genomic Map