Assay Details
| Gene Symbol: HIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY358103.1 | - | 1 | 56 | 63 | |
| JX965370.1 | - | 2 | 224 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
PMS1 homolog 2, mismatch repair system component pseudogene 3 |
PMS2P3 |
PMS2L3, PMS2L9, PMS5, PMSR3 |
-205.3kb | Sense | Chr. 7 - 75507747 - 75528123 on NCBI Build GRCh38 |
| Downstream | C-C motif chemokine ligand 26 | CCL26 |
IMAC, MIP-4a, MIP-4alpha, SCYA26, TSC-1 |
35.7kb | Sense | Chr. 7 - 75769524 - 75791583 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY358103.1 transcripts.
Set Membership: |
|

HIP1
Genomic Map