| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013997.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013997.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600013997.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196395.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196395.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196395.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196395.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196395.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196395.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196395.hg.1 |
TC0600013997.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600196397.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196397.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196397.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196397.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196397.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196397.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196397.hg.1 |
TC0600013997.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013997.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600013997.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013997.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600013997.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196396.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196396.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196396.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196396.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196396.hg.1 |
ENST00000564830.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600196396.hg.1 |
NR_125863.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600196396.hg.1 |
TC0600013997.hg.1 |
Clariom D TC |