Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ENTPD1 antisense RNA 1 | ENTPD1-AS1 | Sense | Chr. 10 - 95753206 - 96090235 on NCBI Build GRCh38 | ||
| Downstream | microRNA 3157 | MIR3157 | mir-3157 | 1.4kb | Sense | Chr. 10 - 96064315 - 96064399 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map