Assay Details
| Gene Symbol: LOC101926997 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK025194.1 | - | 2 | 1551 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 470 | LINC00470 | C18orf2 | -1.2mb | Sense | Chr. 18 - 1268311 - 1359629 on NCBI Build GRCh38 |
| Downstream | methyltransferase like 4 | METTL4 | HsT661 | 19.1kb | Sense | Chr. 18 - 2537525 - 2571503 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC101926997
Genomic Map