Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PLCE1 antisense RNA 2 | PLCE1-AS2 | -142.8kb | Anti-sense | Chr. 10 - 94081950 - 94108794 on NCBI Build GRCh38 | |
| Downstream | PLCE1 antisense RNA 1 | PLCE1-AS1 | 26.0kb | Anti-sense | Chr. 10 - 94279290 - 94287070 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map