Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4497 | MIR4497 | mir-4497 | -34.4kb | Anti-sense | Chr. 12 - 109833348 - 109833436 on NCBI Build GRCh38 |
| Downstream | trichoplein keratin filament binding | TCHP | TpMs | 11.5kb | Anti-sense | Chr. 12 - 109880667 - 109918069 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map