Assay Details
| Gene Symbol: LOC100507551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK125289.1 | - | 2 | 648 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nucleoredoxin-like 1 | NXNL1 |
RDCVF, TXNL6 |
-39.0kb | Sense | Chr. 19 - 17455425 - 17460916 on NCBI Build GRCh38 |
| Downstream | 6-phosphogluconolactonase | PGLS |
6PGL, HEL-S-304 |
10.1kb | Anti-sense | Chr. 19 - 17511623 - 17521291 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK125289.1 transcripts.
This assay is in the 5'UTR region of the AK125289.1 transcripts.
Set Membership: |
|

LOC100507551
Genomic Map